missing translation for 'onlineSavingsMsg'
Learn More

Aminopeptidase P1/XPNPEP1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2970

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447904

  • £349.86 / 100µg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Aminopeptidase P1/XPNPEP1 Polyclonal antibody AF2970 Specifically detects Aminopeptidase P1/XPNPEP1 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Aminopeptidase P1/XPNPEP1
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
NP_065116
Aminoacylproline aminopeptidase, APP1, cytosolic, Cytosolic aminopeptidase P, EC 3.4.11.9, sAmp, xaa-Pro aminopeptidase 1, XPNPEP1, XPNPEPLX-prolyl aminopeptidase (aminopeptidase P)-like, X-prolyl aminopeptidase (aminopeptidase P) 1, soluble, X-prolyl aminopeptidase 1, soluble
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Aminopeptidase P1/XPNPEP1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) XPNPEP2 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
XPNPEP1
E. coli-derived recombinant human Aminopeptidase P1/XPNPEP1 Met1-His623 Accession # NP_065116
Immunogen affinity purified
RUO
Primary
7511
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only