Learn More
Aminopeptidase O/ONPEP Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF4647
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458834
  • £298.33 / 100µg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
Aminopeptidase O/ONPEP Polyclonal antibody AF4647 Specifically detects Aminopeptidase O/ONPEP in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Specifications
Aminopeptidase O/ONPEP
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
Q8N6M6
aminopeptidase O, AOPEP, APO, AP-OFLJ55832, C90RF3, C9orf3, chromosome 9 open reading frame 3, EC 3.4.11.-, FLJ14675, FLJ40923, ONPEP
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Aminopeptidase O/ONPEP in direct ELISAs and Western blots.
Human
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
C9ORF3
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Aminopeptidase O/ONPEP isoform 1 Met1-Phe819 Accession # Q8N6M6
Immunogen affinity purified
RUO
Primary
84909
Reconstitute at 0.2 mg/mL in sterile PBS.