Learn More
Aminopeptidase LRAP/ERAP2 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3830
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448394
  • £433.65 / 100µg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Aminopeptidase LRAP/ERAP2 Polyclonal antibody AF3830 Specifically detects Aminopeptidase LRAP/ERAP2 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Aminopeptidase LRAP/ERAP2
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q6P179
EC 3.4.11, EC 3.4.11.3, endoplasmic reticulum aminopeptidase 2, ERAP2, FLJ23633, FLJ23701, FLJ23807, Leukocyte-derived arginine aminopeptidase, LRAPL-RAPEC 3.4.11.-
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Aminopeptidase LRAP/ERAP2 in direct ELISAs and Western blots.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ERAP2
Mouse myeloma cell line NS0-derived recombinant human Aminopeptidase LRAP/ERAP2 Ala56-Thr960 Accession # Q6P179
Immunogen affinity purified
RUO
Primary
64167
Reconstitute at 0.2 mg/mL in sterile PBS.