missing translation for 'onlineSavingsMsg'
Learn More

AMIGO Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2565

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447644

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

AMIGO Polyclonal antibody AF2565 Specifically detects AMIGO in Human samples. It is validated for Western Blotting
Specifications

Specifications

AMIGO
LYOPH
Western Blot 0.1 ug/mL
Q86WK6
adhesion molecule with Ig-like domain 1, ALI2, Alivin-2, AMIGO1, AMIGO-1, AMIGOamphoterin-induced protein 1, amphoterin-induced gene and ORF, KIAA1163
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human AMIGO in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) AMIGO2 is observed and less than 1% cross-reactivity with rhAMIGO3 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
AMIGO1
Mouse myeloma cell line NS0-derived recombinant human AMIGO Met1-Gly365 Accession # Q86WK6
Antigen Affinity-purified
RUO
Primary
57463
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only