missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMICA/JAML Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand: RD Systems (Bio-Techne) AF3449
Code : Z1
Additional Details : Weight : 0.00010kg
Description
AMICA/JAML Polyclonal antibody AF3449 Specifically detects AMICA/JAML in Human samples. It is validated for ELISA Capture, Flow Cytometry, Western BlottingSpecifications
AMICA/JAML | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Q86YT9 | |
adhesion molecule, interacts with CXADR antigen 1, AMICA, AMICA1, CREA7-1, Gm638, JAML | |
Goat | |
IgG | |
100 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human AMICA/JAML in ELISAs and Western blots. In sandwich ELISAs, less than 0.3% cross-reactivity with recombinant mouse AMICA, recombinant human (rh) JAM-A, rhJAM-B, and rhJAM-C is observed. | |
Human |
Western Blot, Flow Cytometry, CyTOF, ELISA | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
AMICA1 | |
Mouse myeloma cell line NS0-derived recombinant human AMICA/JAML Leu20-Leu275 Accession # Q86YT9 | |
Immunogen affinity purified | |
RUO | |
Primary | |
120425 | |
Reconstitute at 0.2 mg/mL in sterile PBS. |
For Research Use Only