missing translation for 'onlineSavingsMsg'
Learn More

ALK-1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF370

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438324

  • £389.41 / 100µg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ALK-1 Polyclonal antibody AF370 Specifically detects ALK-1 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

ALK-1
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation
P37023
activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRL1, ACVRLK1ORW2, ALK1, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human ALK-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse ALK-1 is observed and less than 1% cross-reactivity with recombinant human (rh) Activin RIA and rhActivin RIB is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ACVRL1
Mouse myeloma cell line NS0-derived recombinant human ALK-1 Asp22-Gln118 Accession # P37023
Immunogen affinity purified
RUO
Primary
94
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only