missing translation for 'onlineSavingsMsg'
Learn More

ALCAM/CD166 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF656

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449804

  • £454.23 / 100µg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ALCAM/CD166 Polyclonal antibody AF656 Specifically detects ALCAM/CD166 in Human samples. It is validated for Blocking (Adhesion), Western Blotting
Specifications

Specifications

ALCAM/CD166
LYOPH
Western Blot 0.1 ug/mL, Adhesion Blockade
AAB59499
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human ALCAM/CD166 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse ALCAM/CD166 is observed.
Human
Western Blot, Adhesion Blockade
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ALCAM
Mouse myeloma cell line NS0-derived recombinant human ALCAM/CD166 Trp28-Ala526 Accession # AAB59499
Immunogen affinity purified
RUO
Primary
214
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only