missing translation for 'onlineSavingsMsg'
Learn More

ADAMTS4 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4307

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13428664

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ADAMTS4 Polyclonal antibody AF4307 Specifically detects ADAMTS4 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

ADAMTS4
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
O75173
a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 4, ADAM metallopeptidase with thrombospondin type 1 motif, 4, ADAM-TS 4, ADAMTS2, ADAMTS-2, ADAM-TS4, ADAMTS-4, ADMP-1, ADMP-1EC 3.4.24.82, Aggrecanase 1, aggrecanase-1, EC 3.4.24, KIAA0688A disintegrin and metalloproteinase with thrombospondin motifs 4
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human ADAMTS4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) ADAMTS5 is observed and less than 1% cross-reactivity with rhADAMTS13, rhADAMTS-L1.2, and rhADAMTS1 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ADAMTS4
Chinese hamster ovary cell line CHO-derived recombinant human ADAMTS4 Phe213-Cys685 Accession # O75173
Immunogen affinity purified
RUO
Primary
9507
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only