missing translation for 'onlineSavingsMsg'
Learn More

ADAM23 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4974

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449014

  • £433.65 / 100µg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ADAM23 Polyclonal antibody AF4974 Specifically detects ADAM23 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

ADAM23
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
O75077
a disintegrin and metalloproteinase domain 23, ADAM metallopeptidase domain 23, disintegrin and metalloproteinase domain-containing protein 23, MDC3, MDC-3, MDC3ADAM 23, MDC-L, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 3
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human ADAM23 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) ADAM8, rhADAM9, and rhADAM22 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ADAM23
Chinese hamster ovary cell line CHO-derived recombinant human ADAM23 Ser60-His585 Accession # O75077
Immunogen affinity purified
RUO
Primary
8745
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only