missing translation for 'onlineSavingsMsg'
Learn More

ADAM22 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4908

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458974

  • £314.17 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

ADAM22 Polyclonal antibody AF4908 Specifically detects ADAM22 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

ADAM22
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q9P0K1
ADAM 22, ADAM metallopeptidase domain 22, MDC2, metalloproteinase-disintegrin ADAM22-3, metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2, MGC149832
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human ADAM22 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ADAM8, rhADAM12, and rhADAM33 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ADAM22
Chinese hamster ovary cell line CHO-derived recombinant human ADAM22 Gly26-His528 (Leu78Val) Accession # Q9P0K1
Immunogen affinity purified
RUO
Primary
53616
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only