missing translation for 'onlineSavingsMsg'
Learn More

Activin C βC subunit Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1629

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487054

  • £377.53 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Activin C βC subunit Polyclonal antibody AF1629 Specifically detects Activin C βC subunit in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Activin C/Inhibin beta C
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
3626
inhibin beta C chain, IHBC, INHBC, inhibin, beta C
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human Activin C beta C subunit in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse Activin C is observed and less than 1% cross-reactivity with recombinant human (rh) Activin A, rhActivin B, rhInhibin A, and rhInhibin B is observed.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
INHBC
E. coli-derived recombinant human Activin C, Gly237-Ser352, Accession # 3626
Antigen Affinity-purified
RUO
Primary
3626
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only