missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTATSF1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518788
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine and rat based on sequence homology.
HTATSF1 Polyclonal antibody specifically detects HTATSF1 in Human samples. It is validated for Western Blot.Specifications
HTATSF1 | |
Polyclonal | |
Unconjugated | |
Htatsf1 | |
1600023H17Rik; 2600017A12Rik; 2700077B20Rik; cofactor required for Tat activation of HIV-1 transcription; dJ196E23.2; HIV TAT specific factor 1; HIV Tat-specific factor 1; HIV Tat-specific factor 1 homolog; HIV-1 Tat specific factor 1; Htatsf1; RGD1563377; RP1-196E23.2; TATSF1; Tat-SF1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
27336 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O43719 | |
Htatsf1 | |
Synthetic peptide sequence (QELYGDGKDGDTQTD) corresponding to the internal amino acids of HTATSF1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |