missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF823 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543572
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GECGEVVLGH SSLNCNIRVD TGHKSCEHQE YGEKPYTHKQ RGKAISHQHS Sequence homology: Human: 100%
ZNF823 Polyclonal antibody specifically detects ZNF823 in Human samples. It is validated for Western Blot.Specifications
ZNF823 | |
Polyclonal | |
Unconjugated | |
ZNF823 | |
HSZFP36; ZFP 36 for a zinc finger protein; ZFP36; Zinc finger protein 823; zinc finger protein ZFP-36; ZNF823 | |
Rabbit | |
Protein A | |
RUO | |
55552 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P16415 | |
ZNF823 | |
Synthetic peptide directed towards the N-terminal of human HSZFP36 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |