missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544330
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: ITITNDKGRL SKDDIDRMVQ EAERYKSEDE ANRDRVAAKN ALESYTYNIK Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 79%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
HSPA2 Polyclonal antibody specifically detects HSPA2 in Human samples. It is validated for Western Blot.Specifications
HSPA2 | |
Polyclonal | |
Unconjugated | |
HSPA2 | |
70 kDa heat shock protein; 70kDa; Hcp70.2; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70kD protein 2; heat shock 70kDa protein 2; Heat shock protein; heat shock protein 2; heat shock protein 70.2; heat shock protein alpha 2; heat shock protein family A (Hsp70) member 2; heat shock protein, 70 kDa 2; heat shock-related 70 kDa protein 2; HSP; HSP70.2; HSP70.3; Hsp70-2; HSP70-3; HSP70A2; Hspa2; Hspt70; HST; Hst70; LOW QUALITY PROTEIN: heat shock-related 70 kDa protein 2; Testis-specific heat shock protein-related; testis-specific heat shock protein-related gene hst70 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3306 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P54652 | |
HSPA2 | |
Synthetic peptide directed towards the middle region of human HSPA2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |