Learn More
HSP60 Mouse anti-Human, Mouse, Rat, Clone: 264233, R&D Systems™
Brand:  RD Systems (Bio-Techne) MAB1800
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458935
  • £271.93 / 100µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
HSP60 Monoclonal antibody MAB1800 Specifically detects HSP60 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Simple Western, Western Blotting
Specifications
Specifications
HSP60
264233
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
HSPD1
E. coli-derived recombinant human HSP60 Met1-Phe573 Accession # P10809
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3329
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunocytochemistry
LYOPH
Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 8-25 ug/mL
P10809
60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HSPD1, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant), SPG13
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human, mouse and rat HSP60.
Human, Mouse, Rat
Videos