missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP40/DNAJB1 Mouse anti-Human, Mouse, Rat, Clone: 419401, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | HSP40/DNAJB1 |
---|---|
Clone | 419401 |
Immunogen | E. coli-derived recombinant human HSP40/DNAJB1 Met1-Ile340 |
Host Species | Mouse |
Primary or Secondary | Primary |
Description
HSP40/DNAJB1 Monoclonal specifically detects HSP40/DNAJB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.Specifications
HSP40/DNAJB1 | |
E. coli-derived recombinant human HSP40/DNAJB1 Met1-Ile340 | |
Primary | |
Human, Mouse, Rat | |
Unconjugated | |
3337 | |
Purified | |
Western Blot | |
RUO |
419401 | |
Mouse | |
Monoclonal | |
IgG2b | |
DnaJ (Hsp40) homolog, subfamily B, member 1, DnaJ (Hsp40) homolog, subfmaily B, member 1, dnaJ homolog subfamily B member 1, DnaJ protein homolog 1, DNAJ1, DNAJB1, hDj-1, HDJ1, Heat shock 40 kDa protein 1, heat shock 40kD protein 1, Heat shock protein 40, HSP40, HSPF1, HSPF1Hdj1, Human DnaJ protein 1, radial spoke 16 homolog B, RSPH16B, Sis1 | |
Supplied as a solution in PBS containing Glycerol and Sodium Azide. with Sodium Azide | |
Protein A or G purified from hybridoma culture supernatant | |
Store the unopened product at -20 to -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |