Learn More
HSP40/DNAJB1 Mouse anti-Human, Mouse, Rat, Clone: 419401, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB4145
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13440426
  • £249.49 / 100µg
Estimated Shipment
28-06-2019
Description
Description
HSP40/DNAJB1 Monoclonal antibody MAB4145 Specifically detects HSP40/DNAJB1 in Human, Mouse, Rat samples. It is validated for Western Blotting
Specifications
Specifications
HSP40/DNAJB1
419401
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DNAJB1
E. coli-derived recombinant human HSP40 Met1-Ile340 Accession # P25685
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3337
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, KnockDown
LYOPH
Western Blot 0.5 ug/mL, Knockout Validated
P25685
DnaJ (Hsp40) homolog, subfamily B, member 1, DnaJ (Hsp40) homolog, subfmaily B, member 1, dnaJ homolog subfamily B member 1, DnaJ protein homolog 1, DNAJ1, DNAJB1, hDj-1, HDJ1, Heat shock 40 kDa protein 1, heat shock 40kD protein 1, Heat shock protein 40, HSP40, HSPF1, HSPF1Hdj1, Human DnaJ protein 1, radial spoke 16 homolog B, RSPH16B, Sis1
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects endogenous human, mouse and rat HSP40 in Western blots.
Human, Mouse, Rat
Videos