missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA512181
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine based on sequence homology.
HSF1 Polyclonal antibody specifically detects HSF1 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
HSF1 | |
Polyclonal | |
Unconjugated | |
Hsf1 | |
AA960185; heat shock factor 1; heat shock factor 2; Heat shock factor protein 1; Heat shock transcription factor 1; heat shock transcription factor 1 alpha isoform; heat shock transcription factor 1 beta isoform; heat shock transcription factor 1 gammaalpha isoform; heat shock transcription factor 1 gammabeta isoform; HSF; HSF 1; Hsf1; Hsf1alpha; Hsf1beta; Hsf1gammaalpha; Hsf1gammabeta; HSTF 1; HSTF1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
15499, 3297 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P38532, Q00613 | |
Hsf1 | |
KLH conjugated synthetic peptide between 278-309 amino acids from human HSF1 Sumoylation Site | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |