missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1C4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529988
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
AKR1C4 Polyclonal antibody specifically detects AKR1C4 in Human samples. It is validated for Western Blot.Specifications
AKR1C4 | |
Polyclonal | |
Unconjugated | |
AKR1C4 | |
3-alpha-HSD; 3-alpha-HSD1; 3-alpha-hydroxysteroid dehydrogenase type I; AKR1C4; Aldo-keto reductase family 1 member C4; aldo-keto reductase family 1, member C4; C11; CDR; CHDR; Chlordecone reductase; chlordecone reductase; 3-alpha hydroxysteroid dehydrogenase, type I; dihydrodiol dehydrogenase 4; DD4; DD-4; Dihydrodiol dehydrogenase 4; dihydrodiol dehydrogenase isozyme DD4; HAKRA; HGNC:387; MGC22581; type I 3-alpha-hydroxysteroid dehydrogenase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1109 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.78 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P17516 | |
AKR1C4 | |
Recombinant protein fragment corresponding to a region within amino acids 37 and 221 of Human HSD3a | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |