missing translation for 'onlineSavingsMsg'
Learn More

HRH1, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA533842

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15372501

  • £314.00 / 50µg


Estimated Shipment
20-06-2018
Add to basket

Description

Description

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey (100%) Marmoset (94%) Orangutan (88%).
Specifications

Specifications

HRH1/H1 Receptor
Polyclonal
Unconjugated
HRH1
Purified
HRH1
Synthetic 17 amino acid peptide from 2nd extracellular domain of human HRH1/Histamine H1 Receptor.
Antigen affinity chromatography
Store at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
3269
Immunohistochemistry (Paraffin)
1mg/mL
PBS with <0.1% sodium azide
P35367
HRH1, H1 histamine receptor, H1 receptor, Histamine 1 receptor, Histamine H1 receptor, H1-R, H1R, Histamine H(1) receptor, Histamine receptor H1, HisH1
Rabbit
IgG
50μg
Antibody
Polyclonal
Human
Videos