HRH1/H1 Receptor Polyclonal Antibody, Invitrogen™

HRH1/H1 Receptor Antibody, PA5-33843, from Invitrogen™. Species Reactivity: Human; Applications: Immunohistochemistry (Paraffin).

Overview
Brand: Invitrogen

Manufacturer Part Number: PA5-33843

UNSPSC: 12352203

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 15312611

Quantity Price
1 £ 314.0 / 50µg
Estimated Shipment date
from Supplier 23-05-2017
Add to basket


Description and Specification

Specification

Antigen HRH1/H1 Receptor
Applications Immunohistochemistry (Paraffin)
Conjugate Unlabeled
Format Purified
Formulation 50μg purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Gene Accession No. P35367
Gene Alias HRH1, H1 histamine receptor, H1 receptor, Histamine 1 receptor, Histamine H1 receptor, H1-R, H1R, Histamine H(1) receptor, Histamine receptor H1, HisH1
Gene Symbols HRH1
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human HRH1/Histamine H1 Receptor.
Isotype IgG
Quantity 50μg
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3269
Target Species Human

Histamine is a ubiquitous messenger molecule released from mast cells, enterochromaffin-like cells, and neurons. Its various actions are mediated by histamine receptors H1, H2, H3 and H4. This gene was thought to be intronless until recently. The protein encoded by this gene is an integral membrane protein and belongs to the G protein-coupled receptor superfamily. It mediates the contraction of smooth muscles, the increase in capillary permeability due to contraction of terminal venules, the release of catecholamine from adrenal medulla, and neurotransmission in the central nervous system. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human HRH1/Histamine H1 Receptor.
Formulation: 50 ug purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Synonyms: HRH1, H1 histamine receptor, H1 receptor, Histamine 1 receptor, Histamine H1 receptor, H1-R, H1R, Histamine H(1) receptor, Histamine receptor H1, HisH1
Uniprot Accession: P35367
Entrez GeneID: 3269
Gene Symbol: HRH1
Percent identity with other species by BLAST analysis: Human (100%) Chimpanzee, Gorilla (95%) Orangutan, Gibbon, Monkey, Marmoset (89%).