Learn More
HPRT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522281
385.63 GBP valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, Molt-4, Raji, mouse liver. Predicted reactivity: Rat (96%), Zebrafish (93%), Xenopus laevis (92%), Dog (96%), Pig (97%), Rabbit (98%), Chicken (94%), Chimpanzee (99%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HPRT1 Polyclonal antibody specifically detects HPRT1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
HPRT1 | |
Polyclonal | |
Unconjugated | |
HPRT1 | |
C81579; HGPRT; HGPRTase; HPGRT; Hprt; HPRT 1; HPRT B; Hprt1; hprt1 protein; hypoxanthine guanine phosphoribosyl transferase; hypoxanthine guanine phosphoribosyl transferase 1; hypoxanthine phosphoribosyl transferase; hypoxanthine phosphoribosyltransferase 1; hypoxanthine-guanine phosphoribosyltransferase; hypoxanthine-guanine phosphoribosyltransferase 1; id:ibd1344; id:ibd5108; testicular tissue protein Li 89; wu:fc10g09; zgc:56221; zgc:86608 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15452, 24465, 3251 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.06 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P00492, P00493, P27605 | |
Hprt, HPRT1 | |
Recombinant fragment corresponding to a region within amino acids 23 and 218 of HPRT (Uniprot ID#P00492) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |