missing translation for 'onlineSavingsMsg'
Learn More

HPRG Rat anti-Mouse, Clone: 242425, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1905

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13429015

  • £388.08 / 500µg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HPRG Monoclonal antibody MAB1905 Specifically detects HPRG in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

HPRG
242425
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
HRG
Mouse myeloma cell line NS0-derived recombinant mouse HPRG Leu19-Lys525 Accession # BAB33094
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3273
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
BAB33094
DKFZp779H1622, Histidine-proline-rich glycoprotein, histidine-rich glycoprotein, HPRGHRGP, HRG
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse HPRG in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 20% cross-reactivity with recombinant human (rh) HPRG is observed and no cross-reactivity with rhCystatin D, F, S, SA, SN, recombinant mouse (rm) Cystatin B, C, E/M, rmKininogen, and rhKininostatin is observed.
Mouse
Videos
Documents

For Research Use Only