missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPi1 Monoclonal Antibody (HIC0-4F9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516126
Code : LD
Additional Details : Weight : 0.10000kg
Description
HPi1 Monoclonal antibody specifically detects HPi1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen).Specifications
HPi1 | |
Monoclonal | |
1 mg/mL | |
tissue culture supernatant with 0.09% sodium azide | |
Mouse | |
100 μL | |
Primary | |
4° C, do not freeze | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
HIC0-4F9 | |
Unconjugated | |
Pancreas Marker HPi1 | |
Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells. | |
RUO | |
Human, Mouse | |
Antibody | |
IgG1 |