missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540887
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: RGPSPGPSGP ANGRHYGIKP ETRAAPAPAT AASTTSSSST SLSSSSKRTE Sequence homology: Cow: 92%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
HOXD9 Polyclonal antibody specifically detects HOXD9 in Human samples. It is validated for Western Blot.Specifications
HOXD9 | |
Polyclonal | |
Unconjugated | |
HOXD9 | |
homeo box D9; homeobox D9; homeobox Hox-D9; homeobox protein Hox-4.4; Homeobox protein Hox-4C; homeobox protein Hox-5.2; Homeobox protein Hox-D9; HOX4; Hox-4.3; Hox-4.3, mouse, homolog of; Hox-4.4; HOX4C; Hox-5.2; HOXD9; Hoxd-9 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3235 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P28356 | |
HOXD9 | |
Synthetic peptide directed towards the middle region of human HOXD9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |