missing translation for 'onlineSavingsMsg'
Learn More

HOXC10, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA535189

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15327604

  • £304.30 / 400µL


Estimated Shipment
07-06-2018
Add to basket

Specifications

Specifications

HOXC10
Polyclonal
PBS with 0.09% sodium azide
Q9NYD6
HOXC10, HOX3I, Homeobox protein Hox-C10, Homeobox protein Hox-3I
Rabbit
IgG
400μL
Antibody
Polyclonal
Human
ImmunocytochemistryþImmunofluorescenceþWestern Blot
Unconjugated
HOXC10
Purified
HOXC10
KLH conjugated synthetic peptide between 152-179 amino acids from the central region of human HOXC10
Antigen affinity chromatography
Store at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
3226
Videos