Learn More
HOXB9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540576
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: SPHASGSLPS VYHPYIQPQG VPPAESRYLR TWLEPAPRGE AAPGQGQAAV Sequence homology: Cow: 100%; Dog: 93%; Horse: 100%; Human: 100%; Mouse: 79%; Pig: 100%; Rabbit: 100%; Rat: 79%
HOXB9 Polyclonal antibody specifically detects HOXB9 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
HOXB9 | |
Polyclonal | |
Unconjugated | |
HOXB9 | |
HGNC:5120; homeo box 2E; homeo box B9; homeobox B9; homeobox protein Hox-2.5; Homeobox protein Hox-2E; homeobox protein Hox-B9; HOX2; HOX-2.5; HOX2E; HOXB9; Hoxb-9; MGC124051; MGC124052; MGC124053; MGC124054 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3219 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P17482 | |
HOXB9 | |
Synthetic peptide directed towards the N-terminal of human HOXB9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |