missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC12 Monoclonal Antibody (3E1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA519125
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: MGEHNLLNPG FVGPLVNIHT GDTFYFPNFR ASGAQLPGLP SLSYPRRDNV CSLSWPSAEP CNGYPQPYLG SPVSLNPPFG RTCELARVED GKGYYREPCA
HOXC12 Monoclonal antibody specifically detects HOXC12 in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
HOXC12 | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
P31275 | |
HOXC12 | |
HOXC12 (NP_776272, 1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunocytochemistry, Western Blot | |
3E1 | |
Unconjugated | |
HOXC12 | |
HOC3F; homeo box C12; homeobox C12; homeobox protein Hox-3.8; Homeobox protein Hox-3F; Homeobox protein Hox-C12; Homeobox protein Hox-C12 (Hox-3F); HOX3; Hox-3.8; HOX3F; HOXC12; Hoxc-12 | |
Mouse | |
Affinity chromatography | |
RUO | |
3228 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |