missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA5 Monoclonal Antibody (2C9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA519104
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: PAQPQIYPWM RKLHISHDNI GGPEGKRART AYTRYQTLEL EKEFHFNRYL TRRRRIEIAH ALCLSERQIK IWFQNRRMKW KKDNKLKSMS MAAAGGAFRP
HOXA5 Monoclonal antibody specifically detects HOXA5 in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
HOXA5 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P20719 | |
HOXA5 | |
HOXA5 (NP_061975.1, 171 a.a. ∽ 270 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Immunocytochemistry, Western Blot | |
2C9 | |
Unconjugated | |
HOXA5 | |
homeo box 1C; homeo box A5; homeobox A5; homeobox protein Hox-1.3; Homeobox protein Hox-1C; homeobox protein HOXA5; homeobox protein Hox-A5; homeobox protein M2; HOX1; HOX1.3; Hox-1.3; HOX1C; Hox-1C; Hoxa5; Hoxa-5; Unknown (protein for MGC:138007) | |
Mouse | |
Affinity chromatography | |
RUO | |
3202 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |