Learn More
hnRNP UL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541832
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: TYPQPSYNQY QQYAQQWNQY YQNQGQWPPY YGNYDYGSYS GNTQGGTSTQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 86%; Rat: 93%
hnRNP UL1 Polyclonal antibody specifically detects hnRNP UL1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
hnRNP UL1 | |
Polyclonal | |
Unconjugated | |
Hnrnpul1 | |
Adenovirus early region 1B-associated protein 5; E130317O14Rik; E1B 55kDa associated protein 5; E1B-55 kDa-associated protein 5; E1BAP5; E1B-AP5; EIB-55kDa associated protein 5; heterogeneous nuclear ribonucleoprotein U like 1; heterogeneous nuclear ribonucleoprotein U-like 1; heterogeneous nuclear ribonucleoprotein U-like protein 1; Hnrnpul; Hnrnpul1; HNRPUL1; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein U-like protein 1 | |
Rabbit | |
Protein A | |
RUO | |
11100 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BUJ2 | |
Hnrnpul1 | |
Synthetic peptide directed towards the C-terminal of human HNRPUL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |