Learn More
hnRNP A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541962
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PGRPQPDSGR RRRRRGEEGH DPKEPEQLRK LFIGGLSFET TDDSLREHFE Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Horse: 90%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
hnRNP A3 Polyclonal antibody specifically detects hnRNP A3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
hnRNP A3 | |
Polyclonal | |
Unconjugated | |
HNRNPA3 | |
2410013L13Rik; 2610209F03Rik; 2610510D13Rik; D10S102; FBRNP; Heterogeneous nuclear ribonucleoprotein A3; hnRNP A3; Hnrnpa3; HNRPA3; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein A3 | |
Rabbit | |
Protein A | |
RUO | |
220988 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P51991 | |
HNRNPA3 | |
Synthetic peptide directed towards the N-terminal of human HNRPA3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |