missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP UL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539206
Code : NEW
Additional Details : Weight : 0.00900kg
Specifications
hnRNP UL2 | |
Polyclonal | |
Unconjugated | |
HNRNPUL2 | |
HNRPUL2, SAF-A2, heterogeneous nuclear ribonucleoprotein U-like protein 2, scaffold-attachment factor A2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221092, 68693 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q1KMD3, Q00PI9 | |
HNRNPUL2 | |
A synthetic peptide derived from the internal region of human HNRNPUL2 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |