missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF4A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526443
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse, rat and Xenopus based on sequence homology.
HNF4A Polyclonal antibody specifically detects HNF4A in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
HNF4A | |
Polyclonal | |
Unconjugated | |
HNF4A | |
alpha transcription factor 4; FLJ39654; FRTS4; hepatic nuclear factor 4 alpha; hepatic nuclear factor 4, alpha; hepatocyte nuclear factor 4 alpha; hepatocyte nuclear factor 4, alpha; hepatocyte nuclear factor 4-alpha; HNF4; Hnf-4; HNF4 alpha; Hnf4a; HNF4a7; HNF4a8; HNF4a9; Hnf4alpha; HNF-4-alpha; HNF4-alpha; HNF4alpha10; HNF4alpha10/11/12; HNF4alpha11; HNF4alpha12; MODY; MODY1; NR2A1; NR2A21; Nuclear receptor 2A1; nuclear receptor subfamily 2 group A member 1; TCF; TCF14; TCF-14; Transcription factor 14; transcription factor HNF-4 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
3172 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P41235 | |
HNF4A | |
KLH conjugated synthetic peptide between 107-139 amino acids from the N-terminal region of human HNF4A | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |