Learn More
HMGB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA116926
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is expected to cross react with porcine, equine and rabbit samples based on 100% sequence homology. Suggested positive control: Hela whole cell extract, antigen standard for HMGB1 (transient overexpression lysate).
HMGB1 Polyclonal antibody specifically detects HMGB1 in Bovine, Canine, Human, Mouse, Ovine, Rabbit, Rat samples. It is validated for ChIP Assay, ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
HMGB1 | |
Polyclonal | |
Unconjugated | |
Hmgb1 | |
Ac2-008; amphoterin; Amphoterin antibody; DEF; DKFZp686A04236; Heparin-binding protein p30; high mobility group 1; high mobility group 1 protein; high mobility group box 1; high mobility group protein 1; High mobility group protein B1; high mobility group protein HMG1; high-mobility group (nonhistone chromosomal) protein 1; high-mobility group box 1; high-mobility-group protein; Hmg1; HMG-1; HMG3; HMG3 antibody; hmgb 1; HMGB1; hmgb-1; HMGB1 protein; hypothetical protein; non-histone protein HMG1; p30; RCJMB04_15a21; RP11-550P23.1; SBP-1; Sulfoglucuronyl carbohydrate binding protein; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100328641, 101112071, 15289, 25459, 282691, 3146, 403170 | |
4° C, do not freeze | |
Liquid |
ChIP Assay, ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
tris glycine with 0.02% sodium azide | |
P09429, P10103, P63158, P63159, Q6YKA4 | |
Hmgb1, LOC101112071 | |
Synthetic peptide between to amino acids 100-200 of the human HMGB1 protein. | |
100 μL | |
Primary | |
Bovine, Canine, Human, Mouse, Ovine, Rabbit, Rat | |
Antibody | |
IgG |