Learn More
HM74A/PUMA-G/GPR109A Rat anti-Human, Clone: 245106, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2760
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449675
  • £390.72 / 500µg
Estimated Shipment date
from Supplier 11-06-2020
Description
Description
HM74A/PUMA-G/GPR109A Monoclonal antibody MAB2760 Specifically detects HM74A/PUMA-G/GPR109A in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
HM74A/PUMA-G/GPR109A/NIACR1
245106
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Q8TDS4
GPR109A, HCA2, HCAR2, hydroxycarboxylic acid receptor 2, Niacr1, Nicotinic Acid Receptor, PUMAG
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human HM74A/GPR109A. Stains human HM74A/GPR109A-transfected cells but not irrelevant transfectants. Cross-reactivity with HM74 (GPR109B) was not tested.
Human
Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
HCAR2
NS0 mouse myeloma cell line transfected with human HM74A/GPR109A Met1-Pro363 Accession # Q8TDS4
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
338442
Reconstitute at 0.5 mg/mL in sterile PBS.