missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR beta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521410
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HLA-DR beta Polyclonal antibody specifically detects HLA-DR beta in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
HLA-DR beta | |
Polyclonal | |
Unconjugated | |
HLA-DRB4 | |
Clone P2-beta-3; DAMC-146E17.2; DR beta-5; DR-16; DR2-beta-2; DR4; DR-4; DR7; DR9; DR-9; DRB1; DRB1 transplantation antigen; DRB4; DRw10; dw2; DW2.2/DR2.2; HLA class II histocompatibility antigen, DR beta 3 chain; HLA class II histocompatibility antigen, DR beta 4 chain; HLA class II histocompatibility antigen, DR beta 5 chain; HLA class II histocompatibility antigen, DR-1 beta chain; HLA class II histocompatibility antigen, DR-5 beta chain; HLA class II histocompatibility antigen, DRB1-15 beta chain; HLA class II histocompatibility antigen, DRB1-16 beta chain; HLA class II histocompatibility antigen, DRB1-3 chain; HLA class II histocompatibility antigen, DRB1-7 beta chain; HLA class II histocompatibility antigen, DRB1-9 beta chain; HLA-DR1B; HLA-DR3B; HLA-DR4B; HLA-DRB; HLA-DRB1; HLA-DRB2; HLA-DRB3; HLA-DRB4; HLA-DRB5; human leucocyte antigen DRB1; human leucocyte antigen DRB3; human leucocyte antigen DRB4; human leucocyte antigen DRB5; leukocyte antigen; leukocyte antigen class II; l | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3123, 3125, 3126, 3127 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P01911, P13762, P79483, Q30154 | |
HLA-DRB1, HLA-DRB3, HLA-DRB4, HLA-DRB5 | |
Recombinant fragment corresponding to a region within amino acids 1 and 266 of HLA DR beta (Uniprot ID#P13762) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |