Learn More
HLA-ABC Mouse anti-Human, NA/LE, Unlabeled, Clone: DX17, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560187
Additional Details : Weight : 0.00880kg
Description
The DX17 monoclonal antibody reacts with a monomorphic epitope expressed on all HLA (human leukocyte antigen) class I molecules examined. DX17 immunoprecipitates HLA class I heavy chains (45kDa) and β2-microglobulin (12kDa) from radiolabeled human cell lines. HLA is determined by a complex segment of the short arm of chromosome 6 and there are many human HLAs encoded in this segment. The antigenic agglomerate is called MHC, for major histocompatibility complex. Examples of class I loci are HLA-A, -B, and -C, which are serologically assayed; class II loci, e.g., HLA-D/DR and DC1, are tested by lymphocytotoxic methods.
Host Species: Mouse
Clone: DX17
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Polyclonal Human NK Cell Line
Blocking Assay, Flow Cytometry
Specifications
HLA-ABC | |
Monoclonal | |
1.0mg/mL | |
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
Polyclonal Human NK Cell Line | |
0.5 mg | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
DX17 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |