missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-A2 Monoclonal Antibody (BB7.2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110344
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes an epitope at the C-terminus of alpha-2 helix and a turn on one of the underlying beta strands within the human HLA-A2 histocompatibility antigen.
HLA-A2 Monoclonal antibody specifically detects HLA-A2 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
HLA-A2 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P04439 | |
HLA-A | |
HLA-A2 solubilised by papain | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunoprecipitation | |
BB7.2 | |
Unconjugated | |
HLA-A | |
A29 protein; antigen presenting molecule; Aw-33; Aw-74; DAQB-90C11.16; FLJ26655; HLA A; HLA class I antigen; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA locus; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLA-DQB1; HLA-DRB1; Human leukocyte antigen A; leucocyte antigen A; leukocyte antigen class I-A; major histocompatibility complex, class I, A; MCH class I antigen; MHC 1; MHC class 1 antigen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen heavy chain; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen null protein; MHC class I antigene; MHC class II antigen; MHC classI antigen; MHC | |
Mouse | |
Protein A | |
RUO | |
3105 | |
4° C, do not freeze | |
Liquid |