Learn More
Pro HGF Activator Rat anti-Mouse, Clone: 198811, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1715
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13428855
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Pro HGF Activator Monoclonal antibody MAB1715 Specifically detects Pro HGF Activator in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
HGF Activator
198811
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
HGFAC
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Pro-HGF Activator Gln35-Ser653 (predicted) Accession # Q9R098
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3083
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9R098
HGF activator, HGFA, Hgfac
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Pro-HGF Activator in Western blots. In Western blots, only the HGFA precursor is recognized. In Western blots, no cross-reactivity with the active form of recombinant mouse HGFA, recombinant human (rh) HGFA, or rhKallikreins 3, 5, and 11 is observed.