missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSV Type 2 gG Monoclonal Antibody (AP1 (B019)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516575
Code : NEW
Additional Details : Weight : 0.01000kg
Description
HSV Type 2 gG Monoclonal antibody specifically detects HSV Type 2 gG in Virus samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
HSV Type 2 gG | |
Monoclonal | |
1 mg/mL | |
PBS with <0.1% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
AP1 (B019) | |
Unconjugated | |
Herpes Simplex 2; Herpes Simplex II; herpes simplex virus 2 gg; HHV-2; HSV Type 2; HSV Type II; HSV2; HSV-2 | |
HSV-2 strain 25766 | |
1 mg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |