missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus Type 1/2 Polyclonal Antibody, FITC, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA127209
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-27209 will not react with HEp-2 cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Herpes Simplex Virus Type 1/2 Polyclonal antibody specifically detects Herpes Simplex Virus Type 1/2 in Virus samples. It is validated for Immunocytochemistry.Specifications
Herpes Simplex Virus Type 1/2 | |
Polyclonal | |
FITC | |
Herpes Simplex 1; Herpes Simplex 2; Herpes Simplex I; Herpes Simplex II; HHV; HHV-2; HSV; HSV 1/2; HSV I/II; HSV Type 1; HSV Type 2; HSV Type II; HSV1; HSV2; HSV-2 | |
Human HSV1, strain F infected cell lysate. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Immunocytochemistry | |
4-5 mg/mL | |
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
Goat | |
IgG fraction | |
RUO | |
Virus | |
Antibody | |
IgG |