missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2 (HER-2) Monoclonal Antibody (N28), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513032
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-13032 targets HER-2 in IP applications and shows reactivity with Human samples. The MA5-13032 immunogen is intact SKBR-3 breast cancer cells.
ErbB2 (HER-2) Monoclonal antibody specifically detects ErbB2 (HER-2) in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation.Specifications
ErbB2 (HER-2) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P04626 | |
ERBB2 | |
Intact SKBR-3 breast cancer cells | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunoprecipitation | |
N28 | |
Unconjugated | |
ERBB2 | |
avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; proto-oncogene c-ErbB-2; Proto-oncogene Neu; receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb | |
Mouse | |
Protein G | |
RUO | |
2064 | |
4° C | |
Liquid |