Learn More
Hepatitis C Virus NS4b Monoclonal Antibody (B29H), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17358
Code : LD
Additional Details : Weight : 0.01000kg
Description
The MA1-7358 antibody reacts with Hepatitis C virus NS4b in viral and infected tissue samples. MA1-7358 has been successfully used in ELISA and Western blot applications. The MA1-7358 antibody was raised against HCV NS4b protein and interacts with the epitope within residues 1689-1735.
Hepatitis C Virus NS4b Monoclonal antibody specifically detects Hepatitis C Virus NS4b in Virus samples. It is validated for ELISA, Western Blot.Specifications
Hepatitis C Virus NS4b | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
B29H | |
Unconjugated | |
HCV; HCV NS4; Hepatitis C Virus; HepC; HepC NS4 | |
NS4b protein. | |
100 μg | |
Primary | |
4° C | |
Liquid |