missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis C Virus Core Antigen Monoclonal Antibody (1F6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17592
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-7592 detects HCV from virus samples. The MA1-7592 immunogen is a purified recombinant HCV core protein.
Hepatitis C Virus Core Antigen Monoclonal antibody specifically detects Hepatitis C Virus Core Antigen in Virus samples. It is validated for ELISA, Western Blot.Specifications
Hepatitis C Virus Core Antigen | |
Monoclonal | |
1 mg/mL | |
PBS with 0.01% sodium azide; pH 7.2 | |
Mouse | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG2a |
ELISA, Western Blot | |
1F6 | |
Unconjugated | |
HCV core; HCVgp1; HepC; HepC core; polyprotein | |
Purified recombinant HCV core protein. | |
100 μg | |
Primary | |
4°C short term, -80°C long term | |
Liquid |