Learn More
Hepatitis B Virus Surface Monoclonal Antibody (T9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513059
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-13059 targets Hepatitis B Virus Surface Antigen in IHC (P) applications and shows reactivity with Human samples. The MA5-13059 immunogen is purified HbsAg, subtype ayw3. Ab-1 recognizes a protein in the outer surface coat of hepatitis B virus.
Hepatitis B Virus Surface Monoclonal antibody specifically detects Hepatitis B Virus Surface in Virus samples. It is validated for Immunohistochemistry (Paraffin).Specifications
Hepatitis B Virus Surface | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
Mouse | |
500 μL | |
Primary | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin) | |
T9 | |
Unconjugated | |
HBsAg; HBV; hepatitis b virus surface antigen; HepB | |
Purified HbsAg, subtype ayw3 | |
RUO | |
Virus | |
Antibody | |
IgG1 λ |