Learn More
Hepatitis B Virus Surface Monoclonal Antibody (E37C), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17347
Code : LD
Additional Details : Weight : 0.01000kg
Description
The MA1-7347 antibody reacts with Hepatitis B virus sAg. capture in viral and infected tissue samples. MA1-7347 has been successfully used in ELISA applications. The MA1-7347 antibody was raised against Hepatitis B Surface antigen.
Hepatitis B Virus Surface Monoclonal antibody specifically detects Hepatitis B Virus Surface in Virus samples. It is validated for ELISA.Specifications
Hepatitis B Virus Surface | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
ELISA | |
E37C | |
Unconjugated | |
HBsAg; HBV; hepatitis b virus surface antigen; HepB | |
Hepatitis B Surface antigen. | |
100 μg | |
Primary | |
4° C | |
Liquid |