Learn More
Hepatitis B Virus Surface Ad/Ay Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA173080
Code : LD
Additional Details : Weight : 0.01000kg
Description
The PA1-73080 antibody reacts with Hepatitis B surface antigen. PA1-73080 has been successfully used in ELISA applications. The PA1-73080 antibody was raised against the surface antigen subtypes ad & ay.
Hepatitis B Virus Surface Ad/Ay Polyclonal antibody specifically detects Hepatitis B Virus Surface Ad/Ay in Virus samples. It is validated for ELISA.Specifications
Hepatitis B Virus Surface Ad/Ay | |
Polyclonal | |
Unconjugated | |
HBsAg; HBV; hepatitis b virus surface antigen; HepB | |
Surface antigen subtypes ad & ay. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
ELISA | |
4-5 mg/mL | |
PBS with 0.1% sodium azide | |
Goat | |
purified | |
RUO | |
Virus | |
Antibody | |
IgG |