missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis B Virus preS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522820
Code : LD
Additional Details : Weight : 0.01000kg
Description
Hepatitis B Virus preS2 Polyclonal antibody specifically detects Hepatitis B Virus preS2 in Virus samples. It is validated for Western Blot.Specifications
Hepatitis B Virus preS2 | |
Polyclonal | |
Unconjugated | |
HBV; HBVgp4; hepatitis b virus pre-s receptor fragment; HepB; pre-s2 | |
Synthetic peptide within residues 130-150 of the 42 kDa preS2 HBV protein. This sequence represents a B cell epitope. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |