missing translation for 'onlineSavingsMsg'
Learn More

Hepassocin Mouse anti-Human, Clone: 198401, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1614

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458775

  • £337.93 / 500µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Hepassocin Monoclonal antibody MAB1614 Specifically detects Hepassocin in Human samples. It is validated for Western Blotting
Specifications

Specifications

Hepassocin/FGL1
198401
Western Blot 1 ug/mL
BAB70690
FGL1, fibrinogen-like 1, fibrinogen-like protein 1, Fibrinogen-like-protein 1, FREP1, Hepassocin, hepatocellular carcinoma-related sequence, Hepatocyte-derived fibrinogen-related protein 1, HFREP-1, HFREP1MGC12455, HP-041, LFIRE1, LFIRE-1, Liver fibrinogen-related protein 1
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Hepassocin in direct ELISAs and Western blots. In direct ELISAs, weak cross-reactivity with rat Hepassocin is observed. In Western blots, 100% cross-reactivity with rat Hepassocin is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FGL1
Mouse myeloma cell line NS0-derived recombinant human Hepassocin Met1-Ile312 Accession # BAB70690
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2267
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only