missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemopexin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529521
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HCT116. Predicted reactivity: Rabbit (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Hemopexin Polyclonal antibody specifically detects Hemopexin in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Specifications
Hemopexin | |
Polyclonal | |
Unconjugated | |
Hpx | |
Beta-1B-glycoprotein; hemopexin; HPX; Hpxn; hx; hyaluronidase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3263 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.89 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P02790 | |
Hpx | |
Recombinant fragment corresponding to a region within amino acids 1 and 234 of Human Hemopexin | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |